Anti-CSTF3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1663AEA 350 EUR
ABGEAT1663A
Anti-CSTF3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CSTF3 (cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR