Anti-TXLNG2P Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1700AEA 350 EUR
ABGEAT1700A
Anti-TXLNG2P Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TXLNG2P (taxilin gamma 2, pseudogene)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR