Anti-DDX43 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1737AEA 350 EUR
ABGEAT1737A
Anti-DDX43 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DDX43 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 43)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR