Anti-DEF6 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1749AEA 350 EUR
ABGEAT1749A
Anti-DEF6 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DEF6 (differentially expressed in FDCP 6 homolog (mouse))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR