Anti-EIF4E2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1877AEA 350 EUR
ABGEAT1877A
Anti-EIF4E2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: EIF4E2 (eukaryotic translation initiation factor 4E family member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR