Anti-EIF4EBP3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1878AEA 350 EUR
ABGEAT1878A
Anti-EIF4EBP3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: EIF4EBP3 (eukaryotic translation initiation factor 4E binding protein 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR