Anti-ELF5 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1896AEA 350 EUR
ABGEAT1896A
Anti-ELF5 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ELF5 (E74-like factor 5 (ets domain transcription factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR