Anti-EMR4P Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1904AEA 350 EUR
ABGEAT1904A
Anti-EMR4P Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: EMR4P (egf-like module containing, mucin-like, hormone receptor-like 4 pseudogene)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR