Anti-ETV1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1952AEA 350 EUR
ABGEAT1952A
Anti-ETV1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ETV1 (ets variant 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR