Anti-FCGR2B Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2027AEA 350 EUR
ABGEAT2027A
Anti-FCGR2B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: FCGR2B (Fc fragment of IgG, low affinity IIb, receptor (CD32))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR