Anti-SRSF10 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2120AEA 350 EUR
ABGEAT2120A
Anti-SRSF10 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SRSF10 (serine/arginine-rich splicing factor 10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR