Anti-FUT10 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2122AEA 350 EUR
ABGEAT2122A
Anti-FUT10 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: FUT10 (fucosyltransferase 10 (alpha (1,3) fucosyltransferase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR