Anti-G3BP1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2137AEA 350 EUR
ABGEAT2137A
Anti-G3BP1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: G3BP1 (GTPase activating protein (SH3 domain) binding protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR