Anti-GALNT6 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2147AEA 350 EUR
ABGEAT2147A
Anti-GALNT6 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GALNT6 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 (GalNAc-T6))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR