Anti-GAPDH Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2152AEA 350 EUR
ABGEAT2152A
Anti-GAPDH Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR