Anti-GCLC Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2176AEA 350 EUR
ABGEAT2176A
Anti-GCLC Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GCLC (glutamate-cysteine ligase, catalytic subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR