Anti-GNAI1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2225AEA 350 EUR
ABGEAT2225A
Anti-GNAI1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GNAI1 (guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR