Anti-GSTM4 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2279AEA 350 EUR
ABGEAT2279A
Anti-GSTM4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GSTM4 (glutathione S-transferase mu 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human