Anti-RBMXL2 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2396AEA 350 EUR
ABGEAT2396A
Anti-RBMXL2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RBMXL2 (RNA binding motif protein, X-linked-like 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human