Anti-HOXB5 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2410AEA 350 EUR
ABGEAT2410A
Anti-HOXB5 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HOXB5 (homeobox B5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human