Anti-HSF4 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2440AEA 350 EUR
ABGEAT2440A
Anti-HSF4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSF4 (heat shock transcription factor 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human