Anti-HSPA1L Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2445AEA 350 EUR
ABGEAT2445A
Anti-HSPA1L Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA1L (heat shock 70kDa protein 1-like)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR