Anti-ID1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2473AEA 350 EUR
ABGEAT2473A
Anti-ID1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ID1 (inhibitor of DNA binding 1, dominant negative helix-loop-helix protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR