Anti-IFNA2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2487AEA 350 EUR
ABGEAT2487A
Anti-IFNA2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IFNA2 (interferon, alpha 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR