Anti-IKBKAP Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2498AEA 350 EUR
ABGEAT2498A
Anti-IKBKAP Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IKBKAP (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR