Anti-IRAK1 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2539AEA 350 EUR
ABGEAT2539A
Anti-IRAK1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IRAK1 (interleukin-1 receptor-associated kinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human