Anti-IRF5 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2547AEA 350 EUR
ABGEAT2547A
Anti-IRF5 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IRF5 (interferon regulatory factor 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR