Anti-ITGB5 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2570AEA 350 EUR
ABGEAT2570A
Anti-ITGB5 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ITGB5 (integrin, beta 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human