Anti-KCNE1 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2592AEA 350 EUR
ABGEAT2592A
Anti-KCNE1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: KCNE1 (potassium voltage-gated channel, Isk-related family, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human