Anti-KCNE1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2593AEA 350 EUR
ABGEAT2593A
Anti-KCNE1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: KCNE1 (potassium voltage-gated channel, Isk-related family, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR