Anti-KRAS Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2651AEA 350 EUR
ABGEAT2651A
Anti-KRAS Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: KRAS (Kirsten rat sarcoma viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR