Anti-LILRA3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2714AEA 350 EUR
ABGEAT2714A
Anti-LILRA3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: LILRA3 (leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR