Anti-MAF Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2761AEA 350 EUR
ABGEAT2761A
Anti-MAF Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MAF (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human