Anti-TAB1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2779AEA 350 EUR
ABGEAT2779A
Anti-TAB1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TAB1 (TGF-beta activated kinase 1/MAP3K7 binding protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR