Anti-MAPK13 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2786AEA 350 EUR
ABGEAT2786A
Anti-MAPK13 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MAPK13 (mitogen-activated protein kinase 13)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human