Anti-MARCH1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2799AEA 350 EUR
ABGEAT2799A
Anti-MARCH1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MARCH1 (membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR