Anti-TADA2B Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2862AEA 350 EUR
ABGEAT2862A
Anti-TADA2B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TADA2B (transcriptional adaptor 2B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human