Anti-MID1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2869AEA 350 EUR
ABGEAT2869A
Anti-MID1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MID1 (midline 1 (Opitz/BBB syndrome))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR