Anti-MPG Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2889AEA 350 EUR
ABGEAT2889A
Anti-MPG Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MPG (N-methylpurine-DNA glycosylase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human