Anti-MPP1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2894AEA 350 EUR
ABGEAT2894A
Anti-MPP1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MPP1 (membrane protein, palmitoylated 1, 55kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR