Anti-MPP3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2895AEA 350 EUR
ABGEAT2895A
Anti-MPP3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MPP3 (membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR