Anti-MRPS27 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2905AEA 350 EUR
ABGEAT2905A
Anti-MRPS27 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MRPS27 (mitochondrial ribosomal protein S27)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human