Anti-MUC4 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2936AEA 350 EUR
ABGEAT2936A
Anti-MUC4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MUC4 (mucin 4, cell surface associated)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human