Anti-NBR1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2978AEA 350 EUR
ABGEAT2978A
Anti-NBR1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NBR1 (neighbor of BRCA1 gene 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR