Anti-NCF4 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT2979AEA 350 EUR
ABGEAT2979A
Anti-NCF4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NCF4 (neutrophil cytosolic factor 4, 40kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human