Anti-NFKB1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3036AEA 350 EUR
ABGEAT3036A
Anti-NFKB1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NFKB1 (nuclear factor of kappa light polypeptide gene enhancer in B-cells 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR