Anti-NKIRAS1 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3054AEA 350 EUR
ABGEAT3054A
Anti-NKIRAS1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NKIRAS1 (NFKB inhibitor interacting Ras-like 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human