Anti-OTUB1 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3158AEA 350 EUR
ABGEAT3158A
Anti-OTUB1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: OTUB1 (OTU deubiquitinase, ubiquitin aldehyde binding 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human