Anti-RPRD1A Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3165AEA 350 EUR
ABGEAT3165A
Anti-RPRD1A Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RPRD1A (regulation of nuclear pre-mRNA domain containing 1A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR