Anti-PAFAH1B1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3169AEA 350 EUR
ABGEAT3169A
Anti-PAFAH1B1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PAFAH1B1 (platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR